Abnova
Manufacturer Code:PAB25864
Catalog # 89-127-493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding Y785 of human ITGB3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ITGB3.
For Research Use Only
Protein Aliases: antigen CD61; CD61; GP3A; GPIIIa; integrin beta 3; Integrin beta-3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa
Gene Aliases: BDPLT16; BDPLT2; CD61; GP3A; GPIIIa; GT; ITGB3
UniProt ID: (Human) P05106
Entrez Gene ID: (Human) 3690
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.