Invitrogen
Manufacturer Code:PA547878
Catalog # PIPA547878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human ICAP-1 Met1-Tyr127 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: bodenin; Bodenin ITGB1BP1 bodenin DKFZp686K08158 ICAP-1 ICAP-1alpha ICAP-1B ICAP1ICAP1AICAP1BICAP-1A integrin beta 1 binding protein 1 integrin beta-1-binding protein 1 Integrin cytoplasmic domain-associated protein 1 integrin cytoplasmic domain-associated protein 1-alpha integrin cytoplasmic domain-associated protein 1-beta; ICAP-1; integrin beta 1 binding protein 1; Integrin beta-1-binding protein 1; Integrin cytoplasmic domain-associated protein 1; integrin cytoplasmic domain-associated protein 1-alpha; integrin cytoplasmic domain-associated protein 1-beta
Gene Aliases: ICAP-1A; ICAP-1alpha; ICAP-1B; ICAP1; ICAP1A; ICAP1B; ITGB1BP1
UniProt ID: (Human) O14713
Entrez Gene ID: (Human) 9270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.