Invitrogen
Manufacturer Code:PA5114752
Catalog # PIPA5114752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ITBP1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI449260 AU019480 Bodenin DKFZp686K08158 Icap1 ICAP-1 ICAP1A ICAP-1A ICAP-1alpha ICAP1B ICAP-1B integrin beta 1 binding protein 1 integrin beta-1-binding protein 1 Integrin cytoplasmic domain-associated protein 1 integrin cytoplasmic domain-associated protein 1-alpha integrin cytoplasmic domain-associated protein 1-beta integrin subunit beta 1 binding protein 1 Itgb1bp1; Bodenin; ICAP-1; integrin beta 1 binding protein 1; Integrin beta-1-binding protein 1; Integrin cytoplasmic domain-associated protein 1; integrin cytoplasmic domain-associated protein 1-alpha; integrin cytoplasmic domain-associated protein 1-beta
Gene Aliases: AI449260; AU019480; ICAP-1A; ICAP-1alpha; ICAP-1B; ICAP1; ICAP1A; ICAP1B; ITGB1BP1
UniProt ID: (Human) O14713, (Mouse) O35671
Entrez Gene ID: (Human) 9270, (Mouse) 16413
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.