Invitrogen
Manufacturer Code:PA567574
Catalog # PIPA567574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human ITGA8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: integrin alpha 8; Integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin, alpha 8; ITGA8 integrin alpha-8 integrin alpha 8
Gene Aliases: ITGA8
UniProt ID: (Human) P53708
Entrez Gene ID: (Human) 8516
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.