Invitrogen
Manufacturer Code:PA547572
Catalog # PIPA547572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse Integrin alpha 8 Phe38-Phe1007 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; ITGA8 integrin alpha-8 integrin alpha 8
Gene Aliases: AI447669; Itga8
UniProt ID: (Mouse) A2ARA8
Entrez Gene ID: (Mouse) 241226
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.