Invitrogen
Manufacturer Code:MA533346
Catalog # PIMA533346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BV3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen CD61; CD49 antigen-like family member E; CD49e; CD61; Fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; GPIIIa; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); Integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; Integrin beta; integrin beta 3; integrin beta 3 (Cd61); Integrin beta-3; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa; very late activation protein 5, alpha subunit; Vitronectin receptor integrin alpha-v/beta-3 cd51/cd61; VLA-5
Gene Aliases: BDPLT16; BDPLT2; CD49e; CD61; FNRA; GP3A; GPIIIa; GT; integrin; ITGA5; Itga5_retired; ITGB3; VLA-5; VLA5A
UniProt ID: (Human) P08648, (Human) P05106, (Rat) Q8R2H2
Entrez Gene ID: (Chicken) 378932, (Human) 3678, (Human) 3690, (Chicken) 374209, (Rat) 315346, (Rat) 29302
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.