Invitrogen
Manufacturer Code:PA533859
Catalog # PIPA533859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from internal region of human ITGA3 / CD49c. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 3 subunit of VLA-3 receptor; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; FRP-2; Galactoprotein B3; GAPB3; integrin alpha 3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); ITGA3 Alpha3 integrin CD49c antigen CD49C GAPB3 Integrin alpha-3 FRP-2 VCA-2 VLA-3 subunit alpha VL3A VLA3a Galactoprotein B3 GAP-B3 ILNEB Integrin alpha3 MSK18; testicular tissue protein Li 96; testicular tissue protein Li 98; very late activation protein 3 receptor, alpha-3 subunit; VLA-3 subunit alpha
Gene Aliases: CD49C; FRP-2; GAP-B3; GAPB3; ILNEB; ITGA3; MSK18; VCA-2; VL3A; VLA3a
UniProt ID: (Human) P26006
Entrez Gene ID: (Human) 3675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.