Invitrogen
Manufacturer Code:PA532880
Catalog # PIPA532880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from extracellular domain of human ITGA2 / CD49b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2 subunit of VLA-2 receptor; BDPLT9 BR CD49B GPIa HPA-5 VLA-2 VLAA2 CD49 antigen-like family member B VLA-2 subunit alpha VLA2 receptor alpha-2 subunit collagen receptor human platelet alloantigen system 5 integrin alpha-2 platelet antigen Br platelet glycoprotein GPIa platelet membrane glycoprotein Ia very late activati; CD49 antigen-like family member B; CD49b; Collagen receptor; GPIa; human platelet alloantigen system 5; Integrin alpha-2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); platelet antigen Br; platelet glycoprotein GPIa; Platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2 subunit alpha
Gene Aliases: BR; CD49B; GPIa; HPA-5; ITGA2; VLA-2; VLAA2
UniProt ID: (Human) P17301
Entrez Gene ID: (Human) 3673
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.