Invitrogen
Manufacturer Code:PA562489
Catalog # PIPA562489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Human ITGA11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Store at &minus20°C long term. Avoid freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT18964 integrin alpha-11; integrin alpha 11; Integrin alpha-11; integrin, alpha 11
Gene Aliases: HsT18964; ITGA11; MSTP018
UniProt ID: (Human) Q9UKX5
Entrez Gene ID: (Human) 22801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.