Invitrogen
Manufacturer Code:PA565539
Catalog # PIPA565539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ITCH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIF4 AIP4 NAPP1 dJ468O1.1 E3 ubiquitin-protein ligase Itchy homolog NFE2-associated polypeptide 1 atrophin-1 interacting protein 4 atrophin-1-interacting protein 4 dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)) itchy homolog E3 ubiquitin protein ligase ubiquitin protein ligase ITCH; AIP4; atrophin-1 interacting protein 4; Atrophin-1-interacting protein 4; E3 ubiquitin-protein ligase Itchy; E3 ubiquitin-protein ligase Itchy homolog; HECT-type E3 ubiquitin transferase Itchy homolog; Itch; itchy E3 ubiquitin protein ligase homolog; NAPP1; NFE2-associated polypeptide 1
Gene Aliases: 6720481N21Rik; 8030492O04Rik; A130065M08; ADMFD; AIF4; AIP4; C230047C07Rik; ITCH; NAPP1
UniProt ID: (Human) Q96J02, (Mouse) Q8C863
Entrez Gene ID: (Human) 83737, (Mouse) 16396
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.