Invitrogen
Manufacturer Code:PA5116631
Catalog # PIPA5116631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 50-150 of human ITCH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6720481N21Rik 8030492O04Rik A130065M08 ADMFD AIF4 AIP4 atrophin-1 interacting protein 4 Atrophin-1-interacting protein 4 C230047C07Rik dJ468O1.1 E3 ubiquitin-protein ligase Itchy E3 ubiquitin-protein ligase Itchy homolog HECT-type E3 ubiquitin transferase Itchy homolog ITCH itch E3 ubiquitin ligase itchy E3 ubiquitin protein ligase itchy E3 ubiquitin protein ligase homolog itchy E3 ubiquitin protein ligase NAPP1 NFE2-associated polypeptide 1; AIP4; atrophin-1 interacting protein 4; Atrophin-1-interacting protein 4; E3 ubiquitin-protein ligase Itchy; E3 ubiquitin-protein ligase Itchy homolog; HECT-type E3 ubiquitin transferase Itchy homolog; Itch; itch E3 ubiquitin ligase; itchy E3 ubiquitin protein ligase homolog; itchy, E3 ubiquitin protein ligase; NAPP1; NFE2-associated polypeptide 1
Gene Aliases: 6720481N21Rik; 8030492O04Rik; A130065M08; ADMFD; AIF4; AIP4; C230047C07Rik; ITCH; NAPP1
UniProt ID: (Human) Q96J02, (Mouse) Q8C863
Entrez Gene ID: (Human) 83737, (Mouse) 16396, (Rat) 311567
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.