Invitrogen
Manufacturer Code:PA552279
Catalog # PIPA552279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ISY1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: functional spliceosome-associated protein 33; ISY1-RAB43 readthrough; Pre-mRNA-splicing factor ISY1 homolog
Gene Aliases: 5830446M03Rik; AI181014; AU020769; FSAP33; ISY1; Isy1-rab43; KIAA1160; RGD1307688
UniProt ID: (Human) Q9ULR0, (Mouse) Q69ZQ2, (Rat) Q6AYB3
Entrez Gene ID: (Human) 57461, (Mouse) 57905, (Rat) 362394
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.