Invitrogen
Manufacturer Code:PA5109868
Catalog # PIPA5109868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-307 of human ISY1 (NP_001186398.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5830446M03Rik AI181014 AU020769 FSAP33 Isy1 Isy1-rab43 KIAA1160 Pre-mRNA-splicing factor ISY1 homolog RGD1307688; functional spliceosome-associated protein 33; Pre-mRNA-splicing factor ISY1 homolog
Gene Aliases: 5830446M03Rik; AI181014; AU020769; FSAP33; ISY1; KIAA1160
UniProt ID: (Human) Q9ULR0, (Mouse) Q69ZQ2
Entrez Gene ID: (Human) 57461, (Mouse) 57905
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.