Invitrogen
Manufacturer Code:PA597212
Catalog # PIPA597212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-335 of human IST1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hIST1; increased sodium tolerance 1 homolog; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; MAPK activating protein PM28; overexpressed in lung cancer 1; Putative MAPK-activating protein PM28
Gene Aliases: 2400003C14Rik; AW536298; IST1; KIAA0174; mKIAA0174; OLC1; RGD1307799
UniProt ID: (Human) P53990, (Rat) Q568Z6, (Mouse) Q9CX00
Entrez Gene ID: (Human) 9798, (Rat) 307833, (Mouse) 71955
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.