Novus Biologicals
Manufacturer Code:NBP2851180.1ML
Catalog # NB033047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human IST1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hIST1; increased sodium tolerance 1 homolog; IST1 homolog; IST1 homolog increased sodium tolerance 1 homolog (yeast) OLC1; IST1, endosomal sorting complex required for transport-III component; overexpressed in lung cancer 1; Putative MAPK-activating protein PM28
Gene Aliases: IST1; KIAA0174; OLC1
UniProt ID: (Human) P53990
Entrez Gene ID: (Human) 9798
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.