Invitrogen
Manufacturer Code:PA525854
Catalog # PIPA525854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 400-429 amino acids from the C-terminal region of human ISPD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog; D-ribitol-5-phosphate cytidylyltransferase; hISPD; Isoprenoid synthase domain-containing protein; Nip 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog isoprenoid synthase domain-containing protein notch1-induced protein hCG&lowbar1745121; notch1-induced protein; testicular tissue protein Li 97
Gene Aliases: 4930579E17Rik; AV040780; CRPPA; hCG_1745121; ISPD; MDDGA7; MDDGC7; Nip
UniProt ID: (Human) A4D126, (Mouse) Q5RJG7
Entrez Gene ID: (Human) 729920, (Mouse) 75847
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.