Novus Biologicals
Manufacturer Code:NBP192030
Catalog # NBP192030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SELNHVKVTSEALGHAGRHLQQIILDQCYNFVCVNVTTSDFQETQKLLSMLEESSLCILYPVVVVSVHFLDFKLVPPSQKMENLMQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog isoprenoid synthase domain containing isoprenoid synthase domain-containing protein; D-ribitol-5-phosphate cytidylyltransferase; hISPD; Isoprenoid synthase domain-containing protein; notch1-induced protein; testicular tissue protein Li 97
Gene Aliases: CRPPA; hCG_1745121; ISPD; MDDGA7; MDDGC7; Nip
UniProt ID: (Human) A4D126
Entrez Gene ID: (Human) 729920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.