Proteintech
Manufacturer Code:186591AP150UL
Catalog # 50-173-2188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ISL2 Fusion Protein Ag13333 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL LIM homeobox 2 ISL2 Islet 2; ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; Islet-2
Gene Aliases: 3110001N10Rik; Isl-2; ISL2
UniProt ID: (Human) Q96A47, (Mouse) Q9CXV0, (Rat) P50480
Entrez Gene ID: (Human) 64843, (Mouse) 104360, (Rat) 57233
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.