Abnova
Manufacturer Code:H00064843K
Catalog # 89-200-583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ISL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ISL2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ10160; Insulin gene enhancer protein ISL-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); Islet-2
Gene Aliases: ISL2
UniProt ID: (Human) Q96A47
Entrez Gene ID: (Human) 64843
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.