Invitrogen
Manufacturer Code:PA530073
Catalog # PIPA530073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 181 of Human ISG20 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD25 HEM45 estrogen-regulated transcript 45 protein interferon-stimulated gene 20 kDa protein promyelocytic leukemia nuclear body-associated protein ISG20; Estrogen-regulated transcript 45 protein; interferon stimulated exonuclease gene 20kDa; Interferon-stimulated gene 20 kDa protein; Promyelocytic leukemia nuclear body-associated protein ISG20
Gene Aliases: CD25; HEM45; ISG20
UniProt ID: (Human) Q96AZ6
Entrez Gene ID: (Human) 3669
Molecular Function: RNA binding protein exoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.