Proteintech
Manufacturer Code:220971AP150UL
Catalog # 50-173-3723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ISG20 Fusion Protein Ag17303 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Estrogen-regulated transcript 45 protein; HEM45 ISG20; interferon stimulated exonuclease; interferon stimulated exonuclease 20; interferon stimulated exonuclease gene 20kDa; interferon stimulated gene 20kDa; Interferon-stimulated gene 20 kDa protein; interferon-stimulated protein; Promyelocytic leukemia nuclear body-associated protein ISG20; Protein DnaQL
Gene Aliases: 1600023I01Rik; 2010107M23Rik; 20kDa; CD25; DnaQL; HEM45; ISG20
UniProt ID: (Human) Q96AZ6, (Mouse) Q9JL16
Entrez Gene ID: (Human) 3669, (Mouse) 57444, (Rat) 293052
Molecular Function:
RNA binding protein
exoribonuclease
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.