Novus Biologicals
Manufacturer Code:NBP271489AF532
Catalog # NP271489A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6C8 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-157 of human ISG15/UCRP (NP_005092) produced in E.coli. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: G1P2interferon-induced 17-kDa/15-kDa protein IFI15hUCRP interferon alpha-inducible protein (clone IFI-15K) Interferon-induced 15 kDa protein Interferon-induced 17 kDa protein interferon-stimulated protein 15 kDa ISG15 ubiquitin-like modifier Ubiquitin cross-reactive protein ubiquitin-like protein ISG15 UCRPIP17; hUCRP; interferon, alpha-inducible protein (clone IFI-15K); Interferon-induced 15 kDa protein; Interferon-induced 17 kDa protein; interferon-induced 17-kDa/15-kDa protein; interferon-stimulated protein, 15 kDa; IP17; Ubiquitin cross-reactive protein; Ubiquitin-like protein ISG15
Gene Aliases: G1P2; hUCRP; IFI15; IMD38; IP17; ISG15; UCRP
UniProt ID: (Human) P05161
Entrez Gene ID: (Human) 9636
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.