Invitrogen
Manufacturer Code:MA529371
Catalog # PIMA529371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 239 |
Immunogen | Recombit Human ISG15 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: hUCRP; interferon, alpha-inducible protein (clone IFI-15K); Interferon-induced 15 kDa protein; Interferon-induced 17 kDa protein; Interferon-induced 17 kDa protein Ubiquitin cross-reactive protein hUCRP Interferon-induced 15 kDa proteinG1P2 UCRP; interferon-induced 17-kDa/15-kDa protein; interferon-stimulated protein, 15 kDa; IP17; Ubiquitin cross-reactive protein; Ubiquitin-like protein ISG15
Gene Aliases: G1P2; hUCRP; IFI15; IMD38; IP17; ISG15; UCRP
UniProt ID: (Human) P05161
Entrez Gene ID: (Human) 9636
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.