Proteintech
Manufacturer Code:148121AP150UL
Catalog # 50-559-474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ISCU Fusion Protein Ag6562 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310020H20Rik HML hnifU ISCU ISU2 NIFU NifU like protein NIFUN; Iron-sulfur cluster assembly enzyme ISCU, mitochondrial; iron-sulfur cluster scaffold homolog; IscU iron-sulfur cluster scaffold homolog; ISCU2; NifU-like N-terminal domain containing; NifU-like N-terminal domain-containing protein; NifU-like protein
Gene Aliases: 2310020H20Rik; AA407971; HML; hnifU; ISCU; ISU2; NIFU; NIFUN; RGD1309562
UniProt ID: (Human) Q9H1K1, (Mouse) Q9D7P6
Entrez Gene ID: (Human) 23479, (Mouse) 66383, (Rat) 288740
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.