Invitrogen
Manufacturer Code:PA571953
Catalog # PIPA571953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 455-483 amino acids from the C-terminal region of human IRX5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeodomain protein IRX-2A; Homeodomain protein IRXB2; Iroquois homeobox protein 5; Iroquois-class homeodomain protein IRX-5; Iroquois-class homeodomain protein IRX-5 Homeodomain protein IRX-2A Homeodomain protein IRXB2 Iroquois homeobox protein 5 IRX5 IRX2A IRXB2
Gene Aliases: HMMS; IRX-2a; IRX2A; IRX5; IRXB2
UniProt ID: (Human) P78411, (Mouse) Q9JKQ4
Entrez Gene ID: (Human) 10265, (Mouse) 54352
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.