Abnova
Manufacturer Code:H00010265A01
Catalog # 89-106-255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 204 a.a. &bsim 264 a.a) partial recombinant protein with GST tag.,IRX5 (NP_005844 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant IRX5.
For Research Use Only
Protein Aliases: Homeodomain protein IRX-2A; Homeodomain protein IRXB2; Iroquois homeobox protein 5; Iroquois-class homeodomain protein IRX-5; IRX-2a; IRXB2
Gene Aliases: HMMS; IRX-2a; IRX2A; IRX5; IRXB2
UniProt ID: (Human) P78411
Entrez Gene ID: (Human) 10265
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.