Invitrogen
Manufacturer Code:PA536256
Catalog # PIPA536256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 200-250 of human IRX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeodomain protein IRXA1; Iroquois homeobox protein 1; Iroquois related homeobox 1; Iroquois-class homeodomain protein IRX-1; Iroquois-class homeodomain protein IRX-1 Homeodomain protein IRXA1 Iroquois homeobox protein 1 IRXA1
Gene Aliases: IRX-5; IRX1; IRXA1
UniProt ID: (Human) P78414, (Mouse) P81068
Entrez Gene ID: (Human) 79192, (Rat) 306659, (Mouse) 16371
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.