Novus Biologicals
Manufacturer Code:NBP2851050.1ML
Catalog # NB033034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human IRX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeodomain protein IRXA1; Homeodomain protein IRXA1 iroquois homeobox 1 Iroquois homeobox protein 1 iroquois-class homeodomain protein IRX-1 IRX-5 IRXA1; Iroquois homeobox protein 1; Iroquois-class homeodomain protein IRX-1
Gene Aliases: IRX-5; IRX1; IRXA1
UniProt ID: (Human) P78414
Entrez Gene ID: (Human) 79192
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.