Invitrogen
Manufacturer Code:MA515072
Catalog # PIMA515072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | J.566.7 |
Immunogen | Synthetic peptide corresponding to the sequence surrounding Ser612 of human IRS-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Insulin receptor substrate 1; IRS-1; pp185
Gene Aliases: G972R; HIRS-1; IRS-1; IRS1; IRS1IRM
UniProt ID: (Human) P35568, (Rat) P35570, (Mouse) P35569
Entrez Gene ID: (Human) 3667, (Rat) 25467, (Mouse) 16367
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.