Novus Biologicals
Manufacturer Code:NBP151966
Catalog # NBP151966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SIHYEGSEYKLSHGS corresponding to internal region according to NP_004127.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: ACO3 EC 4.2.1.3 FLJ23381 IRE-BP 2 Iron regulatory protein 2 iron-responsive element binding protein 2 iron-responsive element-binding protein 2 IRP2IRP2AD; IRE-BP 2; Iron regulatory protein 2; iron-responsive element binding protein 2; Iron-responsive element-binding protein 2; IRP2
Gene Aliases: ACO3; IREB2; IRP2; IRP2AD
UniProt ID: (Human) P48200
Entrez Gene ID: (Human) 3658
Molecular Function:
dehydratase
hydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.