Invitrogen
Manufacturer Code:PA5102893
Catalog # PIPA5102893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IRG1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACOD1 Aconitate decarboxylase aconitate decarboxylase 1 CAD Cis-aconitate decarboxylase cis-aconitic acid decarboxylase Immune-responsive gene 1 protein immune-responsive gene 1 protein homolog immunoresponsive 1 homolog IRG1; Aconitate decarboxylase; Aconitate decarboxylase 1; CAD; Cis-aconitate decarboxylase; Cis-aconitic acid decarboxylase; Immune-responsive gene 1 protein; immune-responsive gene 1 protein homolog; immunoresponsive 1 homolog
Gene Aliases: ACOD1; CAD; IRG1
UniProt ID: (Human) A6NK06
Entrez Gene ID: (Human) 730249
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.