Proteintech
Manufacturer Code:141671AP150UL
Catalog # 50-172-9259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IRF9 Fusion Protein Ag5365 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha-responsive transcription factor subunit; interferon dependent positive acting transcription factor 3 gamma; Interferon regulatory factor 9; interferon regulatory factor 9 IRF 9 IRF9 ISGF 3 gamma ISGF3 ISGF3 p48 subunit ISGF3G p48; Interferon-stimulated gene factor 3 gamma; interferon-stimulated transcription factor 3, gamma (48kD); interferon-stimulated transcription factor 3, gamma 48kDa; IRF-9; ISGF-3 gamma; ISGF3 p48 subunit; Transcriptional regulator ISGF3 subunit gamma
Gene Aliases: IRF-9; IRF9; ISGF3; ISGF3G; p48
UniProt ID: (Human) Q00978, (Mouse) Q61179
Entrez Gene ID: (Human) 10379, (Mouse) 16391, (Rat) 305896
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.