Invitrogen
Manufacturer Code:PA519504
Catalog # PIPA519504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to BSA derived from within residues 450 to the C-terminus of Human IRF5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF-5; SLEB10 IRF-5
Gene Aliases: AW491843; IRF5; mirf5; SLEB10
UniProt ID: (Human) Q13568, (Mouse) P56477
Entrez Gene ID: (Human) 3663, (Rat) 296953, (Mouse) 27056
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.