Invitrogen
Manufacturer Code:711295
Catalog # PI711295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 16 HCLC |
Immunogen | Peptides corresponding to Human IRF4 (aa 425-442 175-192) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: Interferon regulatory factor 4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; Multiple myeloma oncogene 1; NF-EM5
Gene Aliases: IRF4; LSIRF; MUM1; NF-EM5; SHEP8
UniProt ID: (Human) Q15306
Entrez Gene ID: (Human) 3662
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.