Novus Biologicals
Manufacturer Code:NBP178761FR
Catalog # NBP178761FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 200 and 250 of mouse Interferon Regulatory Factor 1 using the numbering given in entry NP_032416.1 (GeneID 16362). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Interferon regulatory factor 1; interferon regulatory factor 1 isoform +I9; interferon regulatory factor 1 isoform d78; interferon regulatory factor 1 isoform delta4; interferon regulatory factor 1 isoform delta7; interferon regulatory factor 1 MARIRF-1; IRF-1
Gene Aliases: IRF-1; IRF1; MAR
UniProt ID: (Human) P10914
Entrez Gene ID: (Human) 3659
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.