Invitrogen
Manufacturer Code:PA541782
Catalog # PIPA541782
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human IREB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IRE-BP 2; Iron regulatory protein 2; iron-regulatory protein 2; iron-responsive element binding protein 2; Iron-responsive element binding protein 2 ACO3 FLJ23381 IRP2 IRP2AD; Iron-responsive element-binding protein 2; Iron-responsive element-binding protein 2-like protein; IRP2
Gene Aliases: ACO3; D9Ertd85e; im:7153062; IREB2; IREBP2; IRP2; IRP2AD
UniProt ID: (Human) P48200, (Rat) Q62751, (Mouse) E9QPM2
Entrez Gene ID: (Human) 3658, (Pig) 100153993, (Dog) 475134, (Horse) 100050493, (Rabbit) 100341900, (Rat) 64831, (Mouse) 64602, (Zebrafish) 652982
Molecular Function:
dehydratase
hydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.