Invitrogen
Manufacturer Code:PA116543
Catalog # APPA6543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminal residues 1-100 of human IRP2 (UniProt P48200). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IRE-BP 2; Iron regulatory protein 2; iron-responsive element binding protein 2; Iron-responsive element binding protein 2 IRE-BP 2 Iron regulatory protein 2; Iron-responsive element-binding protein 2; IRP2
Gene Aliases: ACO3; D9Ertd85e; IREB2; IRP2; IRP2AD
UniProt ID: (Human) P48200, (Mouse) E9QPM2
Entrez Gene ID: (Human) 3658, (Mouse) 64602
Molecular Function:
dehydratase
hydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.