Abnova
Manufacturer Code:PAB15015
Catalog # 89-117-405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 100-200 of mouse Ireb2. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of Ireb2.
For Research Use Only
Protein Aliases: D9Ertd85e; DKFZp564D1164; IRE-BP 2; Iron regulatory protein 2; Iron-responsive element-binding protein 2; IRP2
Gene Aliases: D9Ertd85e; Ireb2; Irp2
UniProt ID: (Mouse) E9QPM2
Entrez Gene ID: (Mouse) 64602
Molecular Function: dehydratase hydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.