Invitrogen
Manufacturer Code:PA522724
Catalog # PIPA522724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide within the human IRE1 alpha protein (within residues 700-800). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum (ER) to nucleus signalling 1; Endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; Inositol-requiring protein 1; IRE1 IRE1P IRE1a hIRE1p ER to nucleus signalling 1 endoplasmic reticulum-to-nucleus signaling 1 inositol-requiring 1 inositol-requiring enzyme 1 inositol-requiring protein 1 ire1-alpha protein kinase/endoribonuclease serine/threonine-protein kinase/endoribonuclease IRE1; Ire1-alpha; IRE1a; protein kinase/endoribonuclease; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1
Gene Aliases: 9030414B18Rik; AI225830; C85377; ERN1; hIRE1p; IRE1; IRE1a; Ire1alpha; IRE1P; RGD1559716
UniProt ID: (Human) O75460, (Mouse) Q9EQY0
Entrez Gene ID: (Human) 2081, (Mouse) 78943, (Rat) 498013
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.