Novus Biologicals
Manufacturer Code:NB1002324F
Catalog # NB1002324F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This IRE1 alpha antibody was raised against a synthetic peptide within the human IRE1 alpha protein (within residues 700-800). [Swiss-Prot #O75460] |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 endoplasmic reticulum to nucleus signaling 1 Endoplasmic reticulum-to-nucleus signaling 1 ER to nucleus signalling 1 FLJ30999 hIRE1p inositol-requiring 1 Inositol-requiring protein 1 IRE 1a IRE1a Ire1-alpha IRE1MGC163279 IRE1P MGC163277 protein kinase/endoribonuclease serine/threonine-protein kinase/endoribonuclease IRE1; Endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; hIRE1p; inositol-requiring 1; inositol-requiring enzyme 1; Inositol-requiring protein 1; Ire1-alpha; IRE1a; protein kinase/endoribonuclease; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1
Gene Aliases: ERN1; hIRE1p; IRE1; IRE1a; IRE1P
UniProt ID: (Human) O75460
Entrez Gene ID: (Human) 2081
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.