Proteintech
Manufacturer Code:182211AP150UL
Catalog # 50-173-2055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IRAK4 Fusion Protein Ag12982 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Interleukin-1 receptor-associated kinase 4; IPD1 IRAK 4 IRAK4 NY REN 64 REN64; IRAK-4; Renal carcinoma antigen NY-REN-64
Gene Aliases: 8430405M07Rik; 9330209D03Rik; IPD1; IRAK-4; IRAK4; NY-REN-64; REN64
UniProt ID: (Human) Q9NWZ3, (Mouse) Q8R4K2
Entrez Gene ID: (Human) 51135, (Mouse) 266632, (Rat) 300177
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.