Novus Biologicals
Manufacturer Code:NBP176782
Catalog # NBP176782
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to 16 amino acids near the carboxy terminus of human IRAK-M. The immunogen is located within the last 50 amino acids of IRAK-M. Amino Acid Squence: CSSKFSWDEYEQYKKE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASRT5 EC 2.7.11.1 FLJ13601 IL-1 receptor-associated kinase M interleukin-1 receptor-associated kinase 3 IRAK-3 IRAKM IRAK-M; IL-1 receptor-associated kinase M; Interleukin-1 receptor-associated kinase 3; IRAK-3; IRAK-M; IRAK3
Gene Aliases: ASRT5; IRAK3; IRAKM
UniProt ID: (Human) Q9Y616
Entrez Gene ID: (Human) 11213
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.