Invitrogen
Manufacturer Code:PA518963
Catalog # PIPA518963
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GHSYGSKPMEKR) corresponding to the internal amino acids of IRAK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-1 receptor-associated kinase M; Interleukin-1 receptor-associated kinase 3; IRAK-3; IRAK-M; irak3; IRAK3 interleukin-1 receptor-associated kinase 3 ASRT5 IRAK-M IRAKM interleukin-1 receptor-associated kinase M
Gene Aliases: 4833428C18Rik; AI563835; IRAK-M; Irak3
UniProt ID: (Mouse) Q8K4B2
Entrez Gene ID: (Mouse) 73914, (Rat) 314870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.