OriGene
Manufacturer Code:TA808811
Catalog # 50-168-2386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-250 of human IP6K2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; IHPK2 PIUS; Inositol hexakisphosphate kinase 2; inositol hexaphosphate kinase 2; InsP6 kinase 2; P(i)-uptake stimulator; pi uptake stimulator; PiUS
Gene Aliases: IHPK2; IP6K2; PIUS; TCCCIA00113
UniProt ID: (Human) Q9UHH9
Entrez Gene ID: (Human) 51447
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.