Invitrogen
Manufacturer Code:PA552449
Catalog # PIPA552449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human IP6K2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; IHPK2 PIUS ATP:1D-myo-inositol-hexakisphosphate phosphotransferase inositol hexaphosphate kinase 2 insp6 kinase 2 pi uptake stimulator; Inositol hexakisphosphate kinase 2; inositol hexaphosphate kinase 2; InsP6 kinase 2; P(i)-uptake stimulator; pi uptake stimulator; PiUS
Gene Aliases: IHPK2; IP6K2; PIUS; TCCCIA00113
UniProt ID: (Human) Q9UHH9
Entrez Gene ID: (Human) 51447
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.