Invitrogen
Manufacturer Code:PA570594
Catalog # PIPA570594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human PPIP5K1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diphosphoinositol pentakisphosphate kinase 1; histidine acid phosphatase domain containing 2A; Histidine acid phosphatase domain-containing protein 2A; hsVIP1; Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1; Inositol pyrophosphate synthase 1; InsP6 and PP-IP5 kinase 1; IP6 kinase; PPIP5K1 HISPPD2A IP6K IPS1 KIAA0377 VIP1; VIP1 homolog
Gene Aliases: HISPPD2A; hsVIP1; IP6K; IPS1; KIAA0377; PPIP5K1; VIP1
UniProt ID: (Human) Q6PFW1
Entrez Gene ID: (Human) 9677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.