Proteintech
Manufacturer Code:105851AP150UL
Catalog # 50-172-6426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | INVS Fusion Protein Ag0934 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: INV inversin INVS KIAA0573 Nephrocystin 2; Inversin; Inversion of embryo turning homolog; inversion of embryo turning protein; inversion of embryonic turning; nephrocystin 2; Nephrocystin-2
Gene Aliases: INV; INVS; NPH2; NPHP2; RGD1563359
UniProt ID: (Human) Q9Y283
Entrez Gene ID: (Human) 27130, (Rat) 313228, (Mouse) 16348
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.