Novus Biologicals
Manufacturer Code:NBP185474
Catalog # NBP185474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LVGQAEPESVLLVHGEAKKMEFLKQKIEQELRVNCYMPANGETVTLPTSPSIPVGISLGLLKREMAQGLLPEAKKPRLLHGTLIMKDSNFRLVSSEQALK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 3-like Cleavage and polyadenylation-specific factor 3-like protein CPSF3-like protein EC 3.1.27 EC 3.1.27.- FLJ20542 INT11 integrator complex subunit 11 INTS11FLJ13294 Protein related to CPSF subunits of 68 kDa RC68 RC-68CPSF73L related to CPSF subunits 68 kDa; Cleavage and polyadenylation-specific factor 3-like protein; CPSF3-like protein; Int11; Integrator complex subunit 11; Protein related to CPSF subunits of 68 kDa; RC-68; related to CPSF subunits 68 kDa
Gene Aliases: CPSF3L; CPSF73L; INT11; INTS11; RC-68; RC68
UniProt ID: (Human) Q5TA45
Entrez Gene ID: (Human) 54973
Molecular Function:
RNA binding protein
endoribonuclease
mRNA polyadenylation factor
mRNA processing factor
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.