Novus Biologicals
Manufacturer Code:NBP266906
Catalog # NBP266906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM22-38 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: EC 2.7.10 EC 2.7.10.1 insulin receptor-related protein insulin receptor-related receptor IRRIR-related receptor; Insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IR-related receptor; IRR
Gene Aliases: INSRR; IRR
UniProt ID: (Human) P14616
Entrez Gene ID: (Human) 3645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.