R&D Systems
Manufacturer Code:AF3055
Catalog # AF3055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human INSRR Leu24-Leu923 Accession # P14616 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 insulin receptor-related protein insulin receptor-related receptor IRR IRRIR-related receptor; Insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IR-related receptor; IRR
Gene Aliases: INSRR; IRR
UniProt ID: (Human) P14616
Entrez Gene ID: (Human) 3645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.